3,5-Dibromo-2-chlorobenzamide

Supplier: Matrix Scientific

063132-500MG
101925-206EA 146.15 USD
101925-206
3,5-Dibromo-2-chlorobenzamide
3,5-Dibromo-2-chlorobenzamide

Hazard: Irritant

Formula: C₇H₄Br₂ClNO
MDL Number: MFCD13187607
CAS Number: 860683-15-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR