1-(3,3-Dimethylbutanoyl)piperidine-4-carboxylic acid

Supplier: Matrix Scientific

063130-500MG
101925-202EA 364.62 USD
101925-202
1-(3,3-Dimethylbutanoyl)piperidine-4-carboxylic acid
1-(3,3-Dimethylbutanoyl)piperidine-4-carboxylic acid

Hazard: Irritant

Formula: C₁₂H₂₁NO₃
MDL Number: MFCD09730776
CAS Number: 953743-57-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR