N-(Methylsulfonyl)alanine

Supplier: Matrix Scientific

063124-500MG
101925-190EA 243.08 USD
101925-190
N-(Methylsulfonyl)alanine
N-(Methylsulfonyl)alanine

Hazard: Irritant

Formula: C₄H₉NO₄S
MDL Number: MFCD07403634
CAS Number: 879123-60-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR