3-(Benzoylamino)-4-methylbenzoic acid

Supplier: Matrix Scientific

Synonyms: 3-Benzamido-4-methylbenzoic acid

063115-500MG
101925-172EA 243.08 USD
101925-172
3-(Benzoylamino)-4-methylbenzoic acid
3-(Benzoylamino)-4-methylbenzoic acid

Hazard: Irritant

Formula: C₁₅H₁₃NO₃
MW: 255.28 g/mol
MDL Number: MFCD00617711
CAS Number: 401828-76-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR