4-(Acetylamino)-5-chloro-2-methoxybenzoic acid
Supplier: Matrix Scientific
063111-500MG
101925-164EA
243.08
USD
101925-164
4-(Acetylamino)-5-chloro-2-methoxybenzoic acid
4-(Acetylamino)-5-chloro-2-methoxybenzoic acid
Hazard: Irritant
Formula:
C₁₀H₁₀ClNO₄ |
MDL Number:
MFCD07783714 CAS Number: 24201-13-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...