4-[(2-Ethylbutanoyl)amino]benzoic acid

Supplier: Matrix Scientific

063104-500MG
101925-150EA 243.08 USD
101925-150
4-[(2-Ethylbutanoyl)amino]benzoic acid
4-[(2-Ethylbutanoyl)amino]benzoic acid

Hazard: Irritant

Formula: C₁₃H₁₇NO₃
MDL Number: MFCD03235452
CAS Number: 486440-98-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR