1,2-Benzisoxazol-6-ol

Supplier: Matrix Scientific

063100-500MG
101925-142EA 307.69 USD
101925-142
1,2-Benzisoxazol-6-ol
1,2-Benzisoxazol-6-ol

Hazard: Irritant

Formula: C₇H₅NO₂
MDL Number: MFCD09999144
CAS Number: 65685-55-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR