4-(Benzoylamino)butanoic acid

Supplier: Matrix Scientific

Synonyms: 4-Benzamidobutanoic acid

063089-500MG
101925-120EA 243.08 USD
101925-120
4-(Benzoylamino)butanoic acid
4-(Benzoylamino)butanoic acid

Hazard: Irritant

Formula: C₁₁H₁₃NO₃
MW: 207.23 g/mol
MDL Number: MFCD00459545
CAS Number: 35340-63-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR