Ethyl-4-[(4-aminophenyl)sulfonyl]piperazine-1-carboxylate

Supplier: Matrix Scientific

063085-500MG
101925-112EA 243.08 USD
101925-112
Ethyl-4-[(4-aminophenyl)sulfonyl]piperazine-1-carboxylate
Ethyl-4-[(4-aminophenyl)sulfonyl]piperazine-1-carboxylate

Hazard: Irritant

Formula: C₁₃H₁₉N₃O₄S
MDL Number: MFCD09271352
CAS Number: 682341-20-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR