4-Amino-N-(3-methoxypropyl)benzenesulfonamide

Supplier: Matrix Scientific

063083-500MG
101925-108EA 364.62 USD
101925-108
4-Amino-N-(3-methoxypropyl)benzenesulfonamide
4-Amino-N-(3-methoxypropyl)benzenesulfonamide

Hazard: Irritant

Formula: C₁₀H₁₆N₂O₃S
MDL Number: MFCD09271351
CAS Number: 27678-19-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR