4-Amino-N,N-dibutylbenzenesulfonamide

Supplier: Matrix Scientific

063080-500MG
101925-102EA 243.08 USD
101925-102
4-Amino-N,N-dibutylbenzenesulfonamide
4-Amino-N,N-dibutylbenzenesulfonamide

Hazard: Irritant

Formula: C₁₄H₂₄N₂O₂S
MDL Number: MFCD04035144
CAS Number: 1709-40-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR