N-[2-(5-Amino-1,3,4-thiadiazol-2-yl)ethyl]-4-bromobenzenesulfonamide

Supplier: Matrix Scientific

063076-500MG
101925-096EA 290.77 USD
101925-096
N-[2-(5-Amino-1,3,4-thiadiazol-2-yl)ethyl]-4-bromobenzenesulfonamide
N-[2-(5-Amino-1,3,4-thiadiazol-2-yl)ethyl]-4-bromobenzenesulfonamide

Hazard: Irritant

Formula: C₁₀H₁₁BrN₄O₂S₂
MDL Number: MFCD13248743
CAS Number: 1199215-87-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR