N-[2-(5-Amino-1,3,4-thiadiazol-2-yl)ethyl]-3,4-dimethoxybenzamide

Supplier: Matrix Scientific

063072-500MG
101925-088EA 364.62 USD
101925-088
N-[2-(5-Amino-1,3,4-thiadiazol-2-yl)ethyl]-3,4-dimethoxybenzamide
N-[2-(5-Amino-1,3,4-thiadiazol-2-yl)ethyl]-3,4-dimethoxybenzamide

Hazard: Irritant

Formula: C₁₃H₁₆N₄O₃S
MDL Number: MFCD13248739
CAS Number: 1199216-01-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR