N-[3-(5-Amino-1,3,4-thiadiazol-2-yl)propyl]-4-methylbenzenesulfonamide

Supplier: Matrix Scientific

063063-500MG
101925-070EA 364.62 USD
101925-070
N-[3-(5-Amino-1,3,4-thiadiazol-2-yl)propyl]-4-methylbenzenesulfonamide
N-[3-(5-Amino-1,3,4-thiadiazol-2-yl)propyl]-4-methylbenzenesulfonamide

Hazard: Irritant

Formula: C₁₂H₁₆N₄O₂S₂
MDL Number: MFCD13248730
CAS Number: 1199216-00-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR