3-Mercapto-5-oxo-4,5-dihydro-1,2,4-triazine-6-carboxylic acid

Supplier: Matrix Scientific

063053-500MG
101925-052EA 484.62 USD
101925-052
3-Mercapto-5-oxo-4,5-dihydro-1,2,4-triazine-6-carboxylic acid
3-Mercapto-5-oxo-4,5-dihydro-1,2,4-triazine-6-carboxylic acid

Hazard: Irritant

Formula: C₄H₃N₃O₃S
MDL Number: MFCD01573074
CAS Number: 7338-75-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR