N-(4-Aminophenyl)-N'-cyclohexylurea

Supplier: Matrix Scientific

063052-500MG
101925-050EA 364.62 USD
101925-050
N-(4-Aminophenyl)-N'-cyclohexylurea
N-(4-Aminophenyl)-N'-cyclohexylurea

Hazard: Irritant

Formula: C₁₃H₁₉N₃O
MDL Number: MFCD01593590
CAS Number: 62972-70-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR