2-(Mesityloxy)acetohydrazide

Supplier: Matrix Scientific

063049-500MG
101925-044EA 243.08 USD
101925-044
2-(Mesityloxy)acetohydrazide
2-(Mesityloxy)acetohydrazide

Hazard: Irritant

Formula: C₁₁H₁₆N₂O₂
MDL Number: MFCD03208905
CAS Number: 347910-45-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR