3-Nitro-1,8-naphthalic anhydride

Supplier: Matrix Scientific

Synonyms: 5-Nitrobenzo[de]isochromene-1,3-dione

063046-500MG
101925-038EA 146.15 USD
101925-038
3-Nitro-1,8-naphthalic anhydride
3-Nitro-1,8-naphthalic anhydride

Hazard: Irritant

Formula: C₁₂H₅NO₅
MW: 243.1718 g/mol
MDL Number: MFCD00006926
CAS Number: 3027-38-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR