1-(4-Ethoxyphenyl)-1H-pyrrole-2,5-dione

Supplier: Matrix Scientific

063039-500MG
101925-024EA 183.08 USD
101925-024
1-(4-Ethoxyphenyl)-1H-pyrrole-2,5-dione
1-(4-Ethoxyphenyl)-1H-pyrrole-2,5-dione

Hazard: Irritant

Formula: C₁₂H₁₁NO₃
MDL Number: MFCD00458572
CAS Number: 19077-60-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR