1-(1-Naphthyl)-1H-pyrrole-2,5-dione

Supplier: Matrix Scientific

063038-500MG
101925-022EA 183.08 USD
101925-022
1-(1-Naphthyl)-1H-pyrrole-2,5-dione
1-(1-Naphthyl)-1H-pyrrole-2,5-dione

Hazard: Irritant

Formula: C₁₄H₉NO₂
MW: 223.23 g/mol
MDL Number: MFCD00019715
CAS Number: 3369-39-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR