2-[(4-Ethoxyphenyl)amino]-4,6-dimethylnicotinamide

Supplier: Matrix Scientific

063034-500MG
101925-014EA 364.62 USD
101925-014
2-[(4-Ethoxyphenyl)amino]-4,6-dimethylnicotinamide
2-[(4-Ethoxyphenyl)amino]-4,6-dimethylnicotinamide

Hazard: Irritant

Formula: C₁₆H₁₉N₃O₂
MDL Number: MFCD01622956
CAS Number: 328548-73-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR