4-Iodo-1H-pyrazole-5-carbohydrazide

Supplier: Matrix Scientific

063033-500MG
101925-012EA 484.62 USD
101925-012
4-Iodo-1H-pyrazole-5-carbohydrazide
4-Iodo-1H-pyrazole-5-carbohydrazide

Hazard: Irritant

Formula: C₄H₅IN₄O
MDL Number: MFCD00605167
CAS Number: 1005592-09-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR