4-Bromo-1H-pyrazole-5-carbohydrazide

Supplier: Matrix Scientific

063032-500MG
101925-010EA 484.62 USD
101925-010
4-Bromo-1H-pyrazole-5-carbohydrazide
4-Bromo-1H-pyrazole-5-carbohydrazide

Hazard: Irritant

Formula: C₄H₅BrN₄O
MDL Number: MFCD00466310
CAS Number: 56976-82-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR