Azobenzene-4,4'-dicarboxylic acid
Supplier: Matrix Scientific
Synonyms:
4,4'-Azodibenzoic Acid
, 4,4'-(Diazene-1,2-diyl)dibenzoic acid
063020-500MG
101924-994EA
304.62
USD
101924-994
Azobenzene-4,4'-dicarboxylic acid
Azobenzene-4,4'-dicarboxylic acid
CAS: 586-91-4
MDL Number: MFCD00496621
Molecular Formula: C14H10N2O4
Molecular Weight: 270,25
Hazard: Irritant
Formula:
C₁₄H₁₀N₂O₄ MW: 270.24 g/mol |
MDL Number:
MFCD00496621 CAS Number: 586-91-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...