9-Oxa-2-azaspiro[5.5]undecane
Supplier: Matrix Scientific
063018-500MG
101924-990EA
484.62
USD
101924-990
9-Oxa-2-azaspiro[5.5]undecane
9-Oxa-2-azaspiro[5.5]undecane
Hazard: Irritant
Formula:
C₉H₁₇NO |
MDL Number:
MFCD12827461 CAS Number: 57289-77-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...