N-[4-(Dimethylamino)benzyl]adamantan-1-amine

Supplier: Matrix Scientific

063011-500MG
101924-976EA 364.62 USD
101924-976
N-[4-(Dimethylamino)benzyl]adamantan-1-amine
N-[4-(Dimethylamino)benzyl]adamantan-1-amine

Hazard: Irritant

Formula: C₁₉H₂₈N₂
MDL Number: MFCD01475821
CAS Number: 289713-75-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR