Ethyl (4-bromo-1H-pyrazol-1-yl)acetate

Supplier: Matrix Scientific

Synonyms: Ethyl 2-(4-bromo-1H-pyrazol-1-yl)acetate

063004-500MG
101924-972EA 364.62 USD
101924-972
Ethyl (4-bromo-1H-pyrazol-1-yl)acetate
Ethyl (4-bromo-1H-pyrazol-1-yl)acetate

Hazard: Irritant

Formula: C₇H₉BrN₂O₂
MW: 233.07 g/mol
MDL Number: MFCD04969156
CAS Number: 82231-58-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR