2,2-Dimethyl-3-piperidin-1-ylpropanal

Supplier: Matrix Scientific

Synonyms: 2,2-Dimethyl-3-(piperidin-1-yl)propanal

063002-500MG
101924-968EA 243.08 USD
101924-968
2,2-Dimethyl-3-piperidin-1-ylpropanal
2,2-Dimethyl-3-piperidin-1-ylpropanal

Hazard: Irritant

Formula: C₁₀H₁₉NO
MW: 169.27 g/mol
MDL Number: MFCD00457347
CAS Number: 37591-27-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR