[1-(1H-Indol-3-yl)ethyl]amine

Supplier: Matrix Scientific

063001-500MG
101924-966EA 606.15 USD
101924-966
[1-(1H-Indol-3-yl)ethyl]amine
[1-(1H-Indol-3-yl)ethyl]amine

Hazard: Irritant

Formula: C₁₀H₁₂N₂
MDL Number: MFCD09965855
CAS Number: 19955-83-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR