5-(1H-Indol-3-yl)-1H-pyrazole-3-carboxylic acid
Supplier: Matrix Scientific
062999-500MG
101924-962EA
552.31
USD
101924-962
5-(1H-Indol-3-yl)-1H-pyrazole-3-carboxylic acid
5-(1H-Indol-3-yl)-1H-pyrazole-3-carboxylic acid
Hazard: Irritant
Formula:
C₁₂H₉N₃O₂ |
MDL Number:
MFCD00441378 CAS Number: 51843-31-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...