3-(1H-Pyrazol-1-yl)propanohydrazide

Supplier: Matrix Scientific

062994-500MG
101924-952EA 364.62 USD
101924-952
3-(1H-Pyrazol-1-yl)propanohydrazide
3-(1H-Pyrazol-1-yl)propanohydrazide

Hazard: Irritant

Formula: C₆H₁₀N₄O
MDL Number: MFCD12030827
CAS Number: 1177300-40-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR