(1Z)-1-(1,3-Thiazol-4-yl)ethanone oxime

Supplier: Matrix Scientific

062985-500MG
101924-934EA 484.62 USD
101924-934
(1Z)-1-(1,3-Thiazol-4-yl)ethanone oxime
(1Z)-1-(1,3-Thiazol-4-yl)ethanone oxime

Hazard: Irritant

Formula: C₅H₆N₂OS
MDL Number: MFCD03550489
CAS Number: 68158-16-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR