Methyl-4-[(methylamino)sulfonyl]benzoate

Supplier: Matrix Scientific

062977-500MG
101924-918EA 221.54 USD
101924-918
Methyl-4-[(methylamino)sulfonyl]benzoate
Methyl-4-[(methylamino)sulfonyl]benzoate

Hazard: Irritant

Formula: C₉H₁₁NO₄S
MDL Number: MFCD09754417
CAS Number: 474398-77-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR