N-(3-Hydroxyphenyl)-2-(4-oxoquinazolin-3(4H)-yl)acetamide

Supplier: Matrix Scientific

062974-500MG
101924-912EA 167.69 USD
101924-912
N-(3-Hydroxyphenyl)-2-(4-oxoquinazolin-3(4H)-yl)acetamide
N-(3-Hydroxyphenyl)-2-(4-oxoquinazolin-3(4H)-yl)acetamide

Hazard: Irritant

Formula: C₁₆H₁₃N₃O₃
MW: 295.3 g/mol
MDL Number: MFCD03714289
CAS Number: 510760-29-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR