Ethyl [(5-ethyl-1,3,4-thiadiazol-2-yl)amino](oxo)acetate

Supplier: Matrix Scientific

062969-500MG
101924-902EA 243.08 USD
101924-902
Ethyl [(5-ethyl-1,3,4-thiadiazol-2-yl)amino](oxo)acetate
Ethyl [(5-ethyl-1,3,4-thiadiazol-2-yl)amino](oxo)acetate

Hazard: Irritant

Formula: C₈H₁₁N₃O₃S
MDL Number: MFCD04278008
CAS Number: 83244-97-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR