2-Chloroethyl[(4-methylphenyl)amino](oxo)acetate

Supplier: Matrix Scientific

062967-500MG
101924-898EA 243.08 USD
101924-898
2-Chloroethyl[(4-methylphenyl)amino](oxo)acetate
2-Chloroethyl[(4-methylphenyl)amino](oxo)acetate

Hazard: Irritant

Formula: C₁₁H₁₂ClNO₃
MDL Number: MFCD13248719
CAS Number: 1192804-63-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR