1-(1,2-Dihydroacenaphthylen-5-yl)-5-oxopyrrolidine-3-carboxylic acid

Supplier: Matrix Scientific

062963-500MG
101924-890EA 364.62 USD
101924-890
1-(1,2-Dihydroacenaphthylen-5-yl)-5-oxopyrrolidine-3-carboxylic acid
1-(1,2-Dihydroacenaphthylen-5-yl)-5-oxopyrrolidine-3-carboxylic acid

Hazard: Irritant

Formula: C₁₇H₁₅NO₃
MDL Number: MFCD03719794
CAS Number: 510737-50-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR