Ethyl-3-(4-chlorophenyl)-1,2,4-oxadiazole-5-carboxylate

Supplier: Matrix Scientific

Synonyms: Ethyl 3-(4-chlorophenyl)-1,2,4-oxadiazole-5-carboxylate

062956-500MG
101924-876EA 364.62 USD
101924-876
Ethyl-3-(4-chlorophenyl)-1,2,4-oxadiazole-5-carboxylate
Ethyl-3-(4-chlorophenyl)-1,2,4-oxadiazole-5-carboxylate

Hazard: Irritant

Formula: C₁₁H₉ClN₂O₃
MW: 252.66 g/mol
MDL Number: MFCD05667194
CAS Number: 163719-69-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR