3-[(1,1-Dioxido-1,2-benzisothiazol-3-yl)amino]phenyl 2-bromobutanoate

Supplier: Matrix Scientific

062943-500MG
101924-850EA 243.08 USD
101924-850
3-[(1,1-Dioxido-1,2-benzisothiazol-3-yl)amino]phenyl 2-bromobutanoate
3-[(1,1-Dioxido-1,2-benzisothiazol-3-yl)amino]phenyl 2-bromobutanoate

Hazard: Irritant

Formula: C₁₇H₁₅BrN₂O₄S
MDL Number: MFCD13248717
CAS Number: 1192805-64-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR