3-[(1,1-Dioxido-1,2-benzisothiazol-3-yl)amino]propyl chloro(phenyl)acetate
Supplier: Matrix Scientific
062942-500MG
101924-848EA
243.08
USD
101924-848
3-[(1,1-Dioxido-1,2-benzisothiazol-3-yl)amino]propyl chloro(phenyl)acetate
3-[(1,1-Dioxido-1,2-benzisothiazol-3-yl)amino]propyl chloro(phenyl)acetate
Hazard: Irritant
Formula:
C₁₈H₁₇ClN₂O₄S |
MDL Number:
MFCD02315022 CAS Number: 356088-58-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...