N-(tert-Butyl)-3-[1-(1,1-dioxido-1,2-benzisothiazol-3-yl)hydrazino]propanamide

Supplier: Matrix Scientific

062939-500MG
101924-842EA 243.08 USD
101924-842
N-(tert-Butyl)-3-[1-(1,1-dioxido-1,2-benzisothiazol-3-yl)hydrazino]propanamide
N-(tert-Butyl)-3-[1-(1,1-dioxido-1,2-benzisothiazol-3-yl)hydrazino]propanamide

Hazard: Irritant

Formula: C₁₄H₂₀N₄O₃S
MDL Number: MFCD01873077
CAS Number: 299923-38-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR