N-(1,1-Dioxido-1,2-benzisothiazol-3-yl)propane-1,3-diamine

Supplier: Matrix Scientific

062934-500MG
101924-832EA 484.62 USD
101924-832
N-(1,1-Dioxido-1,2-benzisothiazol-3-yl)propane-1,3-diamine
N-(1,1-Dioxido-1,2-benzisothiazol-3-yl)propane-1,3-diamine

Hazard: Irritant

Formula: C₁₀H₁₃N₃O₂S
MDL Number: MFCD01442101
CAS Number: 345972-22-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR