3-[(1,1-Dioxido-1,2-benzisothiazol-3-yl)amino]phenol

Supplier: Matrix Scientific

062928-500MG
101924-820EA 364.62 USD
101924-820
3-[(1,1-Dioxido-1,2-benzisothiazol-3-yl)amino]phenol
3-[(1,1-Dioxido-1,2-benzisothiazol-3-yl)amino]phenol

Hazard: Irritant

Formula: C₁₃H₁₀N₂O₃S
MW: 274.3 g/mol
MDL Number: MFCD00763416
CAS Number: 340028-29-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR