(E)-N-(5-Fluoro-4-((hydroxyimino)methyl)-3-iodopyridin-2-yl)pivalamide

Supplier: Matrix Scientific

059528-250MG
101918-334EA 643.08 USD
101918-334
(E)-N-(5-Fluoro-4-((hydroxyimino)methyl)-3-iodopyridin-2-yl)pivalamide
(E)-N-(5-Fluoro-4-((hydroxyimino)methyl)-3-iodopyridin-2-yl)pivalamide

Hazard: Irritant

Formula: C₁₁H₁₃FIN₃O₂
MDL Number: MFCD18803510
CAS Number: 1300019-69-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR