N-(4-Chloro-5-fluoro-3-iodopyridin-2-yl)pivalamide

Supplier: Matrix Scientific

059527-250MG
101918-332EA 643.08 USD
101918-332
N-(4-Chloro-5-fluoro-3-iodopyridin-2-yl)pivalamide
N-(4-Chloro-5-fluoro-3-iodopyridin-2-yl)pivalamide

Hazard: Irritant

Formula: C₁₀H₁₁ClFIN₂O
MDL Number: MFCD18803509
CAS Number: 1299607-56-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR