N-(5-Fluoro-4-formyl-3-iodopyridin-2-yl)pivalamide

Supplier: Matrix Scientific

059510-250MG
101918-298EA 643.08 USD
101918-298
N-(5-Fluoro-4-formyl-3-iodopyridin-2-yl)pivalamide
N-(5-Fluoro-4-formyl-3-iodopyridin-2-yl)pivalamide

Hazard: Irritant

Formula: C₁₁H₁₂FIN₂O₂
MDL Number: MFCD18803495
CAS Number: 1299607-38-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR