(3S,4R)-1-tert-Butyl-3-methyl-4-(8-pivaloyl-5,6,7,8-tetrahydro-1,8-naphthyridin-3-yl)pyrrolidine-1,3-dicarboxylate

Supplier: Matrix Scientific

059504-250MG
101918-286EA 643.08 USD
101918-286
(3S,4R)-1-tert-Butyl-3-methyl-4-(8-pivaloyl-5,6,7,8-tetrahydro-1,8-naphthyridin-3-yl)pyrrolidine-1,3-dicarboxylate
(3S,4R)-1-tert-Butyl-3-methyl-4-(8-pivaloyl-5,6,7,8-tetrahydro-1,8-naphthyridin-3-yl)pyrrolidine-1,3-dicarboxylate

Hazard: Irritant

Formula: C₂₄H₃₅N₃O₅
MDL Number: MFCD18803489
CAS Number: 1420780-19-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR