7-((Trimethylsilyl)ethynyl)-2,3-dihydro[1,4]dioxino[2,3-b]pyridine

Supplier: Matrix Scientific

059503-250MG
101918-284EA 643.08 USD
101918-284
7-((Trimethylsilyl)ethynyl)-2,3-dihydro[1,4]dioxino[2,3-b]pyridine
7-((Trimethylsilyl)ethynyl)-2,3-dihydro[1,4]dioxino[2,3-b]pyridine

Hazard: Irritant

Formula: C₁₂H₁₅NO₂Si
MW: 233.34 g/mol
MDL Number: MFCD18374091
CAS Number: 1261365-41-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR