2,5-Dichloro-3-methoxy-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

Supplier: Matrix Scientific

059501-250MG
101918-280EA 553.85 USD
101918-280
2,5-Dichloro-3-methoxy-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
2,5-Dichloro-3-methoxy-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

Hazard: Irritant

Formula: C₁₂H₁₆BCl₂NO₃
MDL Number: MFCD18803488
CAS Number: 1357387-52-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR