2,3-Dihydro[1,4]dioxino[2,3-b]pyridine-7-carboxylic acid

Supplier: Matrix Scientific

Synonyms: 2,3-Dihydro-[1,4]dioxino[2,3-b]pyridine-7-carboxylic acid

059491-250MG
101918-260EA 643.08 USD
101918-260
2,3-Dihydro[1,4]dioxino[2,3-b]pyridine-7-carboxylic acid
2,3-Dihydro[1,4]dioxino[2,3-b]pyridine-7-carboxylic acid

Hazard: Irritant

Formula: C₈H₇NO₄
MW: 181.15 g/mol
MDL Number: MFCD18250788
CAS Number: 1256818-31-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR